missing translation for 'onlineSavingsMsg'
Learn More

HspA4L Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-35770-100ul

 View more versions of this product

Product Code. 30232251

  • 550.00€ / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

HspA4L Polyclonal antibody specifically detects HspA4L in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HspA4L
Polyclonal
Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
APG1, APG-1, heat shock 70 kDa protein 4L, heat shock 70 kDa protein 4-like protein, heat shock 70kDa protein 4-like, Heat shock 70-related protein APG-1, heat shock protein (hsp110 family), Osmotic stress protein 94, Osp94
Recombinant fusion protein containing a sequence corresponding to amino acids 500-600 of human HspA4L (NP_055093.2).,, Sequence:, KQNLEGDHSDAPMETETSFKNENKDNMDKMQVDQEEGHQKCHAEHTPEEEIDHTGAKTKSAVSDKQDRLNQTLKKGKVKSIDLPIQSSLCRQLGQDLLNSY
100 μL
Primary
Human, Mouse, Rat
Purified
ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
22824
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.