missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ HSPA1B (Human) Recombinant Protein
Human HSPA1B partial ORF ( NP_005336, 531 a.a. - 611 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00003304-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: VQRERVSAKNALESYAFNMKSAVEDEGLKGKISEADKKKVLDKCQEVISWLDANTLAEKDEFEHKRKELEQVCNPIISGLY
Specifications
NP_005336 | |
heat shock 70kDa protein 1B | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HSPA1B | |
GST |
3304 | |
Wheat germ expression system | |
10 μg | |
FLJ54328, HSP70-1B, HSP70-2, HSPA1A | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |