missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ HSPA1B (Human) Recombinant Protein
Human HSPA1B partial ORF ( NP_005336, 531 a.a. - 611 a.a.) recombinant protein with GST-tag at N-terminal.
335.00€ - 508.00€
Specifications
Accession Number | NP_005336 |
---|---|
Gene ID (Entrez) | 3304 |
Name | heat shock 70kDa protein 1B |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16146431
|
Abnova™
H00003304-Q01.10UG |
10 μg |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16156431
|
Abnova™
H00003304-Q01.25UG |
25 μg |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
- Sequence: VQRERVSAKNALESYAFNMKSAVEDEGLKGKISEADKKKVLDKCQEVISWLDANTLAEKDEFEHKRKELEQVCNPIISGLY
Specifications
NP_005336 | |
heat shock 70kDa protein 1B | |
125% SDS-PAGE Stained with Coomassie Blue | |
FLJ54328, HSP70-1B, HSP70-2, HSPA1A | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
3304 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HSPA1B | |
GST |