missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP90 beta Mouse anti-Human, Clone: HSP90AB1/3952, Novus Biologicals™
Mouse Monoclonal Antibody
484.00€
Specifications
| Antigen | HSP90 beta |
|---|---|
| Clone | HSP90AB1/3952 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Peptide Array |
Description
HSP90 beta Monoclonal specifically detects HSP90 beta in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Protein Array.Specifications
| HSP90 beta | |
| 0.2 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Cancer, Cellular Markers, ER Markers, Golgi Apparatus Markers, Hypoxia, Membrane Trafficking and Chaperones, Phospho Specific | |
| 10 mM PBS with 0.05% BSA | |
| 3326 | |
| IgG1 κ | |
| Protein A or G purified |
| HSP90AB1/3952 | |
| Western Blot 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Heat shock 84 kDa, heat shock 90kD protein 1, beta, heat shock 90kDa protein 1, beta, heat shock protein 90kDa alpha (cytosolic), class B member 1, heat shock protein beta, heat shock protein HSP 90-beta, HSP 84, HSP84, HSP90-BETA, HSP90BHSP 90, HSPC2D6S182, HSPCBFLJ26984 | |
| Recombinant human HSP90 beta protein fragment (around aa581-704) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title