missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSP90 beta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
302.00€ - 498.00€
Specifications
| Antigen | HSP90AB1 |
|---|---|
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18233403
|
Novus Biologicals
NBP2-55509 |
100 μL |
498.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18675308
|
Novus Biologicals
NBP2-55509-25ul |
25 μL |
302.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HSP90 beta Polyclonal specifically detects HSP90 beta in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated.Specifications
| HSP90AB1 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cellular Markers, ER Markers, Golgi Apparatus Markers, Hypoxia, Membrane Trafficking and Chaperones | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 3326 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:DEDEVAAEEPNAAVPDEIPPLEGDEDASR | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| D6S182, Heat shock 84 kDa, heat shock 90kD protein 1, beta, heat shock 90kDa protein 1, beta, Heat Shock Protein 90 Alpha Family Class B Member 1, Heat Shock Protein 90 KDa, heat shock protein 90kDa alpha (cytosolic), class B member 1, Heat Shock Protein 90kDa Alpha Family Class B Member 1, heat shock protein HSP 90-beta, HSP 84, HSP 90, HSP84, HSP90B, HSP90-BETA, HSPC2, HSPCB | |
| HSP90AB1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title