missing translation for 'onlineSavingsMsg'
Learn More

HSP27 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB120-5579-0.025mg

 View more versions of this product

Product Code. 18587472

  • 200.00€ / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

HSP27 Polyclonal specifically detects HSP27 in Human, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HSP27
Polyclonal
Unconjugated
Constituents: PBS, 1mg/ml BSA with 0.05% Sodium Azide
HSPB1
Synthetic peptide corresponding to residues L(10) L R G P S W D P F R C(21) of human Heat Shock Protein 27.
0.025 mg
Cancer, Cell Biology, Cell Cycle and Replication, Golgi Apparatus Markers, Membrane Trafficking and Chaperones, Neuroscience
3315
Human
IgG
Western Blot
1 mg/mL
Western Blot 1:1000 - 1:2000, Immunohistochemistry 4 μg/mL, Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:10-1:500
28 kDa heat shock protein, DKFZp586P1322, Estrogen-regulated 24 kDa protein, Heat shock 27 kDa protein, heat shock 27kD protein 1, heat shock 27kDa protein 1, heat shock protein beta-1, HMN2B, HS.76067, Hsp25, HSP27HSP 27, HSP28CMT2F, HspB1, SRP27, Stress-responsive protein 27
Rabbit
Affinity Purified
RUO
Primary
Hsp27
Store at -20°C. Avoid freeze/thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only