missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HSD17B13 Antibody (HSD17B13/13104), Novus Biologicals™
Mouse Monoclonal Antibody
256.00€ - 484.00€
Specifications
| Antigen | HSD17B13 |
|---|---|
| Clone | HSD17B13/13104 |
| Concentration | 0.2 mg/ml |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30295436
|
Novus Biologicals
NBP3-44608-20ug |
20 μg |
256.00€
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30294369
|
Novus Biologicals
NBP3-44608-100ug |
100 μg |
484.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HSD17B13 Monoclonal antibody specifically detects HSD17B13 in Human samples. It is validated for nanSpecifications
| HSD17B13 | |
| 0.2 mg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| HMFN0376, hydroxysteroid (17-beta) dehydrogenase 13, member 3,17-beta-hydroxysteroid dehydrogenase 13, MGC138508,17-beta-HSD 13, NIIL497, SCDR9, short-chain dehydrogenase/reductase 9, UNQ497/PRO1014 | |
| Recombinant fragment (around aa1-200) of human HSD17B13 protein (Uniprot: Q7Z5P4). Exact sequence is proprietary. | |
| Primary | |
| Store at 4⪚C. |
| HSD17B13/13104 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Endocrinology, Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| 345275 | |
| IgG2b κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title