missing translation for 'onlineSavingsMsg'
Learn More

HROB Antibody (7A3A9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication

Brand:  Novus Biologicals NBP2-37407-0.025ml

 View more versions of this product

Product Code. 18620786

  • 234.00€ / 0.03mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

HROB Monoclonal specifically detects HROB in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HROB
Monoclonal
1 mg/mL
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:50, CyTOF-ready
chromosome 17 open reading frame 53, FLJ11594, hypothetical protein LOC78995, MCM8IP, MGC3130
Mouse
69.8 kDa
0.025 mL
Primary
Human
IgG2b
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, CyTOF
7A3A9
Unconjugated
PBS with 0.05% Sodium Azide
C17orf53
Purified recombinant fragment of human C17ORF53 (AA: 282-527) expressed in E. Coli.
Affinity Purified
RUO
78995
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only