missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HRH3 Monoclonal Antibody (1008944)
GREENER_CHOICE

Product Code. 30535779
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30535779 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30535779 Supplier Invitrogen™ Supplier No. MA560857

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Reconstitute at 0.5 mg/mL in sterile PBS.

H3 Histamine Receptor regulates the release of the neurotransmitters norepinephrine and somatostatin. In histamine-containing neurons of the brain, the H3 receptor acts as a presynaptic autoreceptor, while in nonhistamine-containing neurons, in both the central and peripheral nervous systems, it acts as a presynaptic heteroreceptor. Multiple isoforms of H3 receptors are produced by alternative splicing. Histamine H3 receptor has been reported primarily in brain. ESTs have been isolated from brain (normal and cancer), breast cancer, colon cancer, and retinoblastoma libraries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HRH3
Applications Flow Cytometry
Classification Monoclonal
Clone 1008944
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with trehalose and no preservative
Gene HRH3
Gene Accession No. Q9Y5N1
Gene Alias AW049250; G protein-coupled receptor; G protein-coupled receptor 97; GPCR97; G-protein coupled receptor 97; H3R; HH3R; histamine H3 receptor; histamine H3 receptor H3L isoform; histamine H3 receptor H3S isoform; histamine receptor; histamine receptor H 3; histamine receptor H3; Hrh3
Gene Symbols HRH3
Host Species Mouse
Immunogen Synthetic peptide containing human Histamine H3 R.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11255
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.