missing translation for 'onlineSavingsMsg'
Learn More

HPV18 E6 Mouse anti-Virus, Clone: HPV18/1297, Novus Biologicals™

Mouse Monoclonal Antibody

484.00€

Specifications

Antigen HPV18 E6
Clone HPV18/1297
Concentration 0.2 mg/mL
Dilution Immunohistochemistry-Paraffin 1 to 2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18656956
View Documents
Novus Biologicals
NBP3-07521-100ug
100 μg
484.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

HPV18 E6 Monoclonal specifically detects HPV18 E6 in Virus samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

HPV18 E6
0.2 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Virus
E6, HpV18gp1, Human papillomavirus type 18 E6, Protein E6
IgG1 κ
Protein A or G purified
HPV18/1297
Immunohistochemistry-Paraffin 1 to 2 μg/mL
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
HPV18 E6-beta-galactosidase fusion protein
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.