missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPV18 E6 Antibody (C1N1), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54807H
This item is not returnable.
View return policy
Description
HPV18 E6 Monoclonal antibody specifically detects HPV18 E6 in Virus samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
| HPV18 E6 | |
| Monoclonal | |
| HRP | |
| E6, HpV18gp1, Human papillomavirus type 18 E6, Protein E6 | |
| HPV18E6-beta-galactosidase fusion protein synthesized in E. coli | |
| 0.1 mL | |
| Cancer, Tumor Suppressors | |
| Virus | |
| Purified |
| Western Blot, ELISA, Immunoprecipitation | |
| C1N1 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction