missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPV16 E7 Antibody (716-332cc), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
519.00€
Specifications
| Antigen | HPV16 E7 |
|---|---|
| Clone | 716-332cc |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Alexa Fluor 532 |
Description
HPV16 E7 Monoclonal antibody specifically detects HPV16 E7 in Virus samples. It is validated for Western Blot, ELISASpecifications
| HPV16 E7 | |
| Western Blot | |
| Alexa Fluor 532 | |
| Mouse | |
| Virus | |
| E7, HPV16 E7, HpV16gp2, Links Transforming protein human papillomavirus 16, Protein E7, Transforming protein, Transforming protein human papillomavirus 16 | |
| IgG2a | |
| Protein A purified |
| 716-332cc | |
| Monoclonal | |
| Purified | |
| RUO | |
| 50mM Sodium Borate | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with HPV oncoprotein E7, type 16. | |
| Primary | |
| Store at 4°C in the dark. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title