Learn More
Protein kinase Y linked Antibody (OTI3E10), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44632
This item is not returnable.
View return policy
Description
Protein kinase Y linked Monoclonal antibody specifically detects Protein kinase Y linked in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Protein kinase Y linked | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized powder ( 1X PBS, pH 7.3,) | |
| Mouse | |
| >95% | |
| RUO | |
| Primary | |
| For reconstitution, we recommend adding 100uL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiment, we strongly recommend performing another round of desalting process. | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Lyophilized |
| Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| OTI3E10 | |
| Western Blot 1:500, Immunohistochemistry 1:50, Immunocytochemistry/ Immunofluorescence 1:100 | |
| EC 2.7.11, EC 2.7.11.1, protein kinase, Y-linked | |
| Full-length protein expressed in 293T cell transfected with human PRKY expression vector | |
| 100 μg | |
| Protein Kinase, Wnt Signaling Pathway | |
| 5616 | |
| Human | |
| Antibody | |
| IgG2a |