missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXA2 Monoclonal Antibody (OTI6A6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812718
This item is not returnable.
View return policy
Description
Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.Specifications
| HOXA2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O43364 | |
| HOXA2 | |
| Human recombit protein fragment corresponding to amino acids 57-319 of human HOXA2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI6A6 | |
| Unconjugated | |
| HOXA2 | |
| HOX1K, MCOHI | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3199 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction