missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOXA2 Monoclonal Antibody (OTI6A6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812718S
This item is not returnable.
View return policy
Description
HOXA2 Monoclonal antibody specifically detects HOXA2 in Human, Rat samples. It is validated for Western BlotSpecifications
| HOXA2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8WW01 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 116461, 289083 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI6A6 | |
| Unconjugated | |
| Tsen15 | |
| AI324701; homeo box A2; homeobox A2; homeobox protein Hox-1.11; Homeobox protein Hox-1K; homeobox protein Hox-A2; Hox1.11; Hox-1.11; Hox111; Hox-111; HOX1K; HOXA2; Hoxa-2; MCOHI; NEWGENE_2813 | |
| Full length human recombinant protein of human TSEN15 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human, Rat | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction