missing translation for 'onlineSavingsMsg'
Learn More

Hormone-sensitive Lipase/HSL Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications

178.00€ - 423.00€

Specifications

Antigen Hormone-sensitive Lipase/HSL
Dilution Western Blot 1:100-1:2000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18715053
View Documents
Novus Biologicals
NB110-37253SS
0.025 mL
178.00€
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18770924
View Documents
Novus Biologicals
NB110-37253
0.05 mL
423.00€
0.05mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Hormone-sensitive Lipase/HSL Polyclonal specifically detects Hormone-sensitive Lipase/HSL in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

Hormone-sensitive Lipase/HSL
Western Blot
Unconjugated
Rabbit
Cancer, Cholesterol Metabolism, Diabetes Research, Lipid and Metabolism, Lipid Droplets, Signal Transduction
PBS containing 1 mg/ml BSA with 0.05% Sodium Azide
3991
A fusion protein containing an internal portion of the rat HSL protein (between residues 699-908). [Swiss-Prot P15304]
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Western Blot 1:100-1:2000
Polyclonal
Purified
RUO
Human, Mouse, Rat
hormone-sensitive lipase, hormone-sensitive lipase testicular isoform, HSLEC 3.1.1.79, LHS, lipase, hormone-sensitive
LIPE
IgG
IgG purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.