missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abcam HORAC Assay Kit, Hydroxyl Radical Measurement, 192 Tests
Description
- Measure hydroxyl radical
- Provided in a 192-test format
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Format | Kit |
| No. of Reactions | 192 Tests |
| Target Molecule | Hydroxyl Radical |
| Detection Method | Fluorescence |
| Quantity | 192 Tests |
| For Use With (Application) | Hydroxyl radical measurement |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?