Learn More
HOMER2 Monoclonal Antibody (OTI2D5), TrueMAB™, OriGene
Product Code. 17586433
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17586433 30 μL 30µL
17566433 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17586433 Supplier Origene Technologies Supplier No. TA806310S
This item requires a price update prior to being ordered.
Request a Price for this item here.
Mouse Monoclonal Antibody
HOMER2 Monoclonal antibody specifically detects HOMER2 in Human samples. It is validated for Western Blot
Specifications
Antigen HOMER2
Applications Western Blot
Classification Monoclonal
Clone OTI2D5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Mrrf
Gene Accession No. Q96E11
Gene Alias 9330120H11Rik; A VESL-2; ACPD; AW539445; CPD; Cupidin; DFNA68; homer 2; homer homolog 2; homer homolog 2 (Drosophila); homer homolog 3; homer protein homolog 2; homer scaffold protein 2; homer scaffolding protein 2; homer, neuronal immediate early gene, 2; HOMER2; homer-2; VASP/Ena-related gene up-regulated during seizure and LTP 2; Vesl2; Vesl-2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 56-262 of human MRRF produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92399
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a