missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hnRNP L Antibody (4D11), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-6106AF532
This item is not returnable.
View return policy
Description
hnRNP L Monoclonal specifically detects hnRNP L in Human, Mouse, Rat, Amphibian, Bovine, Chicken, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications
| hnRNP L | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| HNRNPL | |
| Human hnRNP proteins from HeLa cells purified by affinity chromatography on ssDNA agarose. | |
| Primary | |
| Human, Mouse, Rat, Amphibian, Bovine, Chicken, Primate | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| 4D11 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
| FLJ35509, heterogeneous nuclear ribonucleoprotein L, hnRNP L, hnRNP-L, HNRPL | |
| Mouse | |
| 0.1 mL | |
| 3191 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction