missing translation for 'onlineSavingsMsg'
Learn More

hnRNP L Antibody (4D11), Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18239233 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18239233 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18239233 Supplier Novus Biologicals Supplier No. NB1206106AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

hnRNP L Monoclonal specifically detects hnRNP L in Human, Mouse, Rat, Amphibian, Bovine, Chicken, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen hnRNP L
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 4D11
Conjugate Alexa Fluor 350
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias FLJ35509, heterogeneous nuclear ribonucleoprotein L, hnRNP L, hnRNP-L, HNRPL
Gene Symbols HNRNPL
Host Species Mouse
Immunogen Human hnRNP proteins from HeLa cells purified by affinity chromatography on ssDNA agarose.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3191
Target Species Human, Mouse, Rat, Amphibian, Bovine, Chicken, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.