missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNF-3 alpha/FoxA1 Antibody (3B2), Novus Biologicals™
Rabbit Monoclonal Antibody
343.00€ - 533.00€
Specifications
| Antigen | HNF-3 alpha/FoxA1 |
|---|---|
| Clone | 3B2 |
| Dilution | Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:200-1:1000 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043213
|
Novus Biologicals
NBP3-26572-50ul |
50 μL |
343.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043290
|
Novus Biologicals
NBP3-26572-100ul |
100 μL |
533.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HNF-3 alpha/FoxA1 Monoclonal antibody specifically detects HNF-3 alpha/FoxA1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, ImmunoprecipitationSpecifications
| HNF-3 alpha/FoxA1 | |
| Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:200-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| forkhead box A1, Forkhead box protein A1, hepatocyte nuclear factor 3, alpha, hepatocyte nuclear factor 3-alpha, HNF-3A, HNF-3-alpha, HNF3ATCF-3A, MGC33105, TCF3A, Transcription factor 3A | |
| A synthesized peptide derived from Human HNF-3 alpha/FoxA1 [UniProt P55317] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 3B2 | |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer, Stem Cell Markers | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 3169 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title