missing translation for 'onlineSavingsMsg'
Learn More

HMGXB4 Monoclonal Antibody (OTI2D3), TrueMAB™, OriGene

Product Code. 17590373
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17590373 30 μL 30µL
17570373 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17590373 Supplier Origene Technologies Supplier No. TA801928S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HMGXB4 Monoclonal antibody specifically detects HMGXB4 in Human samples. It is validated for Western Blot

Specifications

Antigen HMGXB4
Applications Western Blot
Classification Monoclonal
Clone OTI2D3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PIK3CD
Gene Accession No. O00329
Gene Alias high mobility group protein 2-like 1; high-mobility group protein 2-like 1; HMG box domain containing 4; HMG box-containing protein 4; HMG domain-containing protein 4; HMG2L1; HMGBCG; HMG-box containing 4; HMGXB4; Protein HMGBCG; Protein THC211630; THC211630
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 286-610 of human PIK3CD produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5293
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.