missing translation for 'onlineSavingsMsg'
Learn More

HMGB4 Mouse anti-Human, Clone: OTI5G3, liquid, TrueMAB™

Product Code. 17550064
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17550064 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17550064 Supplier Origene Technologies Supplier No. TA808611

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HMGB4 contains two HMG-box regions, which is found in a variety of eukaryotic chromosomal proteins and transcription.

Specifications

Antigen HMGB4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI5G3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene HMGB4
Gene Accession No. Q8WW32
Gene Alias dJ1007G16.5
Gene Symbols HMGB4
Host Species Mouse
Immunogen Full length human recombit protein of human HMGB4 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127540
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.