missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ HLA-E (Human) Recombinant Protein

Human HLA-E full-length ORF (no protein_acc, 1 a.a. - 358 a.a.) recombinant protein with GST tag at N-terminal.

Brand:  Abnova™ H00003133-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16106191

  • 528.00€ / 25µg

Estimated Shipment: 04-07-2023
to see stock.
Add to basket

Description

Description

  • Sequence: MVDGTLLLLLSEALALTQTWAGSHSLKYFHTSVSRPGRGEPRFISVGYVDDTQFVRFDNDAASPRMVPRAPWMEQEGSEYWDRETRSARDTAQIFRVNLRTLRGYYNQSEAGSHTLQWMHGCELGPDRRFLRGYEQFAYDGKDYLTLNEDLRSWTAVDTAAQISEQKSNDASEAEHQRAYLEDTCVEWLHKYLEKGKETLLHLEPPKTHVTHHPISDHEATLRCWALGFYPAEITLTWQQDGEGHTQDTELVETRPAGDGTFQKWAAVVVPSGEEQRYTCHVQHEGLPEPVTLRWKQASQPTIPIVGIIAGLVLLGSVVSGAVVAAVIWRKKSSGGKGGSYSKAEWSDSAQGSESHSL
Specifications

Specifications

3133
Wheat germ expression system
25 μg
DKFZp686P19218, EA1.2, EA2.1, HLA-6.2, MHC, QA1
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
major histocompatibility complex, class I, E
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HLA-E
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers