missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DRA Antibody (1C11A5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61863
This item is not returnable.
View return policy
Description
HLA DRA Monoclonal specifically detects HLA DRA in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| HLA DRA | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:500 | |
| FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MLRW | |
| Mouse | |
| 58.8 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1C11A5 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| HLA-DRA | |
| Purified recombinant fragment of human HLA-DRA (AA: 26-254) expressed in E. Coli. | |
| Protein G purified | |
| Adaptive Immunity, Cell Biology, Diabetes Research, Immunology | |
| 3122 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction