missing translation for 'onlineSavingsMsg'
Learn More

HLA-DR, Mouse anti-Human, APC-C750, Clone: GRB-1, Abnova™

Product Code. 16063576
Change view
Click to view available options
Quantity:
100 Reactions
Unit Size:
100 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16063576 100 Reactions 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16063576 Supplier Abnova Supplier No. MAB15428.100Reactions

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specifications

Antigen HLA-DR
Applications Flow Cytometry
Classification Monoclonal
Clone GRB-1
Conjugate APC-C750
Dilution Flow Cytometry (20 uL/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
Host Species Mouse
Immunogen Mononuclear cell leukemia acute undifferentiated.
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store in the dark at 4°C. Avoid prolonged exposure to light.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.