missing translation for 'onlineSavingsMsg'
Learn More

HLA DOB Antibody (DOB 10E6/5), Biotin, Novus Biologicals™

Product Code. 18146858 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18146858 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18146858 Supplier Novus Biologicals Supplier No. NBP250420B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HLA DOB Monoclonal specifically detects HLA DOB in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA DOB
Applications Western Blot, ELISA
Classification Monoclonal
Clone DOB 10E6/5
Conjugate Biotin
Dilution Western Blot, ELISA
Formulation PBS with 0.05% Sodium Azide
Gene Alias DOB, FLJ57033, HLA class II histocompatibility antigen, DO beta chain, major histocompatibility complex, class II, DO beta, MHC class II antigen DOB
Gene Symbols HLA-DOB
Host Species Mouse
Immunogen C-terminal peptide MHC class II
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Asthma, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3112
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.