missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA Class I Antibody (W6/32) - Low Endotoxin, Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-09091
This item is not returnable.
View return policy
Description
HLA Class I Monoclonal specifically detects HLA Class I in Human, Bovine, Feline, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Functional, CyTOF-ready.
Specifications
| HLA Class I | |
| Monoclonal | |
| 1 mg/ml | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Functional, CyTOF-ready | |
| A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80 | |
| Membrane of human tonsil cells | |
| 0.1 mg | |
| Adaptive Immunity, Cell Biology, Immunology | |
| 3105 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay, CyTOF | |
| W6/32 | |
| Unconjugated | |
| PBS, 0.2 um filter sterilized | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Bovine, Feline, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction