Learn More
HLA class I ABC Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15240-1-AP-150UL
This item is not returnable.
View return policy
Description
HLA-class I major histocompatibility (MHC) antigens are intrinsic membrane glycoproteins expressed on nucleated cells and noncovalently associated with an invariant beta2 microglobulin. They carry foreign determits important for immune recognition by cytotoxic T cells, thus important for anti-viral and anti-tumour defence. Human HLA-class I antigens are represented by HLA-A, HLA-B and HLA-C molecules.Specifications
| HLA class I ABC | |
| Polyclonal | |
| Unconjugated | |
| HLA-A | |
| HLA A, HLA class I, HLA class I (HLA-A), HLA class I ABC, HLA-A, MHC class I antigen A*3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3105 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P30443 | |
| HLA-A | |
| HLA class I ABC Fusion Protein Ag7370 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |