missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA B Antibody (2G7B10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
236.00€ - 608.00€
Specifications
| Antigen | HLA B |
|---|---|
| Clone | 2G7B10 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18212954
|
Novus Biologicals
NBP2-61871 |
0.1 mL |
608.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18618678
|
Novus Biologicals
NBP2-61871-0.025ml |
0.025 mL |
236.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HLA B Monoclonal specifically detects HLA B in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| HLA B | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| Adaptive Immunity, Immunology | |
| HLA-B | |
| IgG1 | |
| Protein G purified |
| 2G7B10 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Unconjugated | |
| Mouse | |
| 3106 | |
| Purified recombinant fragment of human HLA-B (AA: 241-362) expressed in E. Coli. | |
| Primary | |
| 58.8 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title