missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA A2 Antibody (BB7.2) - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-43553
This item is not returnable.
View return policy
Description
HLA A2 Monoclonal antibody specifically detects HLA A2 in Human samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| HLA A2 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A *1, MHC class I antigen A *11, MHC class I antigen A *80 | |
| The original antibody was generated by immunizing BALB/c mice with papain solubilized HLA A and B7 antigens. (Uniprot# P04439) | |
| 0.2 mg | |
| Adaptive Immunity, Cell Biology, Immunology | |
| 3105 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry, ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| BB7.2 | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction