Learn More
HJ 753-2015 Pyrethroid Pesticides Mixture 611 1000 μg/mL in Acetone, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | (±)-Fenpropathrin∣Allethrin∣Bifenthrin∣Cypermethrin∣Deltamethrin∣Fenvalerate∣lambda-Cyhalothrin∣Tetramethrin |
| CAS | 39515-41-8, 584-79-2, 82657-04-3, 52315-07-8, 52918-63-5, 51630-58-1 |
| Analyte or Component Names | (±)-Fenpropathrin∣Allethrin∣Bifenthrin∣Cypermethrin∣Deltamethrin∣Fenvalerate∣lambda-Cyhalothrin∣Tetramethrin |
| Shipping Condition | +20°C |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetone |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | (±)-Fenpropathrin 1000 μg/mL∣Allethrin 1000 μg/mL∣Bifenthrin 1000 μg/mL∣Cypermethrin 1000 μg/mL∣Deltamethrin 1000 μg/mL∣Fenvalerate 1000 μg/mL∣lambda-Cyhalothrin 1000 μg/mL∣Tetramethrin 1000 μg/mL |