missing translation for 'onlineSavingsMsg'
Learn More

HJ 753-2015 Pyrethroid Pesticides Mixture 153 100 μg/mL in Acetone, Dr. Ehrenstorfer

Product Code. 30310817
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30310817 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30310817 Supplier Dr. Ehrenstorfer Supplier No. DREA50000153AC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (±)-Fenpropathrin∣Allethrin∣Bifenthrin∣Cypermethrin∣Deltamethrin∣Fenvalerate∣lambda-Cyhalothrin∣Tetramethrin
CAS 39515-41-8, 584-79-2, 82657-04-3, 52315-07-8, 52918-63-5, 51630-58-1
Analyte or Component Names (±)-Fenpropathrin∣Allethrin∣Bifenthrin∣Cypermethrin∣Deltamethrin∣Fenvalerate∣lambda-Cyhalothrin∣Tetramethrin
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Acetone
Quantity 1 mL
Concentration or Composition (by Analyte or Components) (±)-Fenpropathrin 100 μg/mL∣Allethrin 100 μg/mL∣Bifenthrin 100 μg/mL∣Cypermethrin 100 μg/mL∣Deltamethrin 100 μg/mL∣Fenvalerate 100 μg/mL∣lambda-Cyhalothrin 100 μg/mL∣Tetramethrin 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.