Learn More
HJ 683-2014 Aldehyde/Ketone-DNPH Mixture 622 3 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Click to view available options
Quantity:
1.2 mL
Unit Size:
1.2mL
Specifications
Specifications
| CAS | 958-60-1, 1019-57-4, 1567-89-1, 888-54-0, 1157-84-2, 1527-98-6 |
| Quantity | 1.2 mL |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetonitrile |