Learn More
IBA Lifesciences™ HIV p24-Gag Peptide KAFSPEVIPMF (HLA-B*5701)
Shop All IBA Lifesciences Products
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
HIV p24-Gag peptide KAFSPEVIPMF (HLA-B*5701) is a single peptide for stimulation of T cells. The peptide from human immunodeficiency virus 1 (HIV-1) p24 group-specific antigen (Gag) is synthesized as it is presented by the MHC class I HLA-B*5701 allele and can be used for targeted in vitro generation and expansion of antigen-specific CD8+ T cells.
- Peptides have been purified by HPLC using a water and acetonitrile gradient
- Endotoxin-free
Specifications
Specifications
| Purity | >85% |
| Endotoxin Level | Endotoxin-free |
| For Use With (Application) | T Cell Stimulation |
| Lyophilized | Yes |
| Quantity | 1 mg |