Learn More
HIV-1, gp120, Mouse anti-Viruses, Clone: 1G12, Abnova™
Mouse Monoclonal Antibody
Brand:  Abnova MAB0036-M08.50ug
Product Code. 16079715
  • 410.00€ / 50µg
Specifications
HIV-1, gp120
Monoclonal
Unconjugated
In PBS, pH 7.4
Recombinant Fc fusion protein corresponding to amino acids 255-477 of human Immunodeficiency Virus type 1 (HIV-1), CN54 gp120 outer domain.
RUO
Human
Antibody
IgG1 κ
ELISA, Western Blot
1G12
The optimal working dilution should be determined by the end user.
Mouse
50 μg
Primary
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.