missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H4, ac Lys16 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB21-2077
This item is not returnable.
View return policy
Description
Histone H4 Polyclonal specifically detects Histone H4 in Human, Mouse, Rat, Porcine, Mammal samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Dot Blot, Chromatin Immunoprecipitation (Negative).
Specifications
| Histone H4 (ac Lys16) | |
| Polyclonal | |
| Unconjugated | |
| PBS and 30% Glycerol with 0.05% Sodium Azide | |
| H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, H4K16ac, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, histone 4, H4, histone cluster 4, H4, histone H4, MGC24116 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 121504 | |
| Human, Mouse, Rat, Pig, Mammalia | |
| IgG |
| ChIP Assay, Dot Blot, Immunocytochemistry, Immunofluorescence | |
| 0.62 mg/mL | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100, Dot Blot 1:1000, Chromatin Immunoprecipitation (Negative) | |
| P62805, P62806 | |
| HIST4H4 | |
| Synthetic acetylated peptide surrounding Lysine 16 of human Histone H4 [Swiss Prot P62805]. | |
| 0.05 mg | |
| Primary | |
| Human, mouse and rat. Predicted to react with most mammal species. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction