missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Target Protein: Histone H3
- Form: Liquid
- Species Reactivity: Human
- Purity: >90%
- Tested Applications: BL
- Composition: pH 7.4; 0.87% Sodium chloride, 0.476% HEPES, 0.0584% EDTA, 0.001% Sorbitan monolaurate, ethoxylated
- Usage: Synthetic blocking peptide
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Product Type | Blocker and Diluent |
| Buffer | 0.476% HEPES, 0.87% sodium chloride, 0.0584% EDTA, 0.001% sorbitan monolaurate, ethoxylated, pH7.4 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -20°C. |
| Form | Liquid |
| Label or Dye | Unconjugated |
| Reactivity | Human |
| Shipping Condition | Blue Ice |
| Substrate Properties | Peptide-based Substrate |
| Target Molecule | Histone H3 (di methyl K9) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?