missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abcam Histone H3 (K4) Methyltransferase Activity Quantification Assay Kit, 48 Tests
Shop All Abcam ProductsDescription
- Applications: Enzyme activity assay
- Sample types: Tissue Extracts, Cell Lysate
- Reactive species: Mouse, Human
- Assay time: 3 hr
- Results type: Quantitative
- Detection method: Microplate reader
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Product Type | Western Blot Kit |
| For Use With (Equipment) | Microplate Reader |
| Format | Kit |
| No. of Reactions | 48 Tests |
| Reactivity | Mouse, Human |
| Run Time | 3 hr. |
| Target Molecule | Histone H3 (K4) Methyltransferase |
| Detection Method | Colorimetric |
| Cross Reactivity | Mouse, Human |
| Sample Type | Tissue Extracts, Cell Lysates |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?