missing translation for 'onlineSavingsMsg'
Learn More

Histone H3.1, Dimethyl Lys9 Antibody (1A10), Novus Biologicals™

Rabbit Monoclonal Antibody

343.00€ - 533.00€

Specifications

Antigen Histone H3.1
Clone 1A10
Dilution Western Blot 1:500-1:5000, ELISA, Immunocytochemistry/Immunofluorescence 1:30-1:200
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30043240
View Documents
Novus Biologicals
NBP3-26621-50ul
50 μL
343.00€
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30043615
View Documents
Novus Biologicals
NBP3-26621-100ul
100 μL
533.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Histone H3.1 Monoclonal antibody specifically detects Histone H3.1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry
Specifications

Specifications

Histone H3.1
Western Blot 1:500-1:5000, ELISA, Immunocytochemistry/Immunofluorescence 1:30-1:200
Monoclonal
Purified
RUO
Human
H3 histone family, member D, H3.1HIST1H3D, H3/d, H3FA, H3FB, H3FC, H3FDHIST1H3F, H3FF, H3FH, H3FI, H3FJ, H3FK, H3FL, HIST1H3A, HIST1H3B, HIST1H3C, HIST1H3G, HIST1H3H, HIST1H3I, HIST1H3J, histone 1, H3e, histone cluster 1, H3e, histone H3.1, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h, Histone H3/i, Histone H3/j, Histone H3/k, Histone H3/l
Synthesized peptide [UniProt P68431]
Primary
Store at -20 to -70C. Avoid freeze-thaw cycles.
1A10
Western Blot, ELISA, Immunocytochemistry
Unconjugated
Rabbit
Epigenetics, Phospho Specific
PBS, pH 7.4, 150mM NaCl and 50% glycerol
8353
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.