missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2AY/macroH2A.1 Antibody (CL5256) [CoraFluor™ 1], Novus Biologicals™
Shop All Bio Techne ProductsDescription
Histone H2AY/macroH2A.1 Monoclonal antibody specifically detects Histone H2AY/macroH2A.1 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Histone H2AY/macroH2A.1 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | CL5256 |
| Conjugate | CoraFluor 1 |
| Formulation | PBS |
| Gene Alias | core histone macro-H2A.1, H2A histone family, member Y, H2A.y, H2A/y, H2AF12M, H2AFJ, Histone H2A.y, Histone macroH2A1, histone macroH2A1.1, histone macroH2A1.2, MACROH2A1, MACROH2A1.1, macroH2A1.2, Medulloblastoma antigen MU-MB-50.205, mH2A1 |
| Host Species | Mouse |
| Immunogen | This antibody was developed using a synthetic peptide derived from O75367, with the exact immunogen sequence remaining proprietary. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?