missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Histone H2AY/macroH2A.1 Antibody (CL5256) [Alexa Fluor™ 594], Novus Biologicals™
Shop alle Bio Techne produkterBeskrivelse
Histone H2AY/macroH2A.1 Monoclonal antibody specifically detects Histone H2AY/macroH2A.1 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Tekniske data
Tekniske data
| Antigen | Histone H2AY/macroH2A.1 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | CL5256 |
| Conjugate | Alexa Fluor 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | core histone macro-H2A.1, H2A histone family, member Y, H2A.y, H2A/y, H2AF12M, H2AFJ, Histone H2A.y, Histone macroH2A1, histone macroH2A1.1, histone macroH2A1.2, MACROH2A1, MACROH2A1.1, macroH2A1.2, Medulloblastoma antigen MU-MB-50.205, mH2A1 |
| Host Species | Mouse |
| Immunogen | This antibody was developed using a synthetic peptide derived from O75367, with the exact immunogen sequence remaining proprietary. |
| Purification Method | Protein A purified |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?