Learn More
Histone H2AX, p Ser139 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 25 publications
Brand: Novus Biologicals NB100-2280
This item is not returnable.
View return policy
Description
Histone H2AX (p Ser139) Polyclonal antibody specifically detects Histone H2AX (p Ser139) in Human, Mouse, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications
| Histone H2AX (p Ser139) | |
| Polyclonal | |
| RUO |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| This Histone H2AX [p Ser139] Antibody was developed against a synthetic phospho-peptide, which represented a portion of the C-terminus of human histone H2AX surrounding phosphorylated serine 139 (GeneID 3014). | |
| Store at 4C. Do not freeze. |