Learn More
Histone H2AX, p Ser139 Antibody, Novus Biologicals™
Product Code. 18758323 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18758323 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18758323 Supplier Novus Biologicals Supplier No. NB100384
Rabbit Polyclonal Antibody
Histone H2AX (p Ser139) Polyclonal antibody specifically detects Histone H2AX (p Ser139) in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), ChIP assay, Immunoassay.
TRUSTED_SUSTAINABILITY
Specifications
Antigen Histone H2AX (p Ser139)
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:10000-1:25000, Simple Western 5 ug/ml, Flow Cytometry 5 ug per 1 million cells, Immunohistochemistry 1:2000 - 1:10000, Immunocytochemistry/Immunofluorescence 1:500 to 1:5000, Immunohistochemistry-Paraffin 1:2000 - 1:10000, Immunohistochemistry-Frozen 1:1000 - 1:5000, Chromatin Immunoprecipitation (ChIP), Knockout Validated
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. P16104
Gene Alias H2A.X, H2A/X, H2AFX
Gene Symbols H2AFX
Host Species Rabbit
Immunogen This Histone H2AX [p Ser139] Antibody was developed against to a region surrounding phosphorylated serine 139 of human histone H2AX [Swiss-Prot entry P16104] (GeneID 3014).
Molecular Weight of Antigen 15 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Mitotic Regulators, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3014
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Rhesus Monkey, Gorilla, Crab-eating macaque, Thirteen-lined ground squirrel and Little brown bat.
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4C. Do not freeze.
Isotype IgG