missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2a Rabbit anti-Yeast, Clone: S08-1A5, Novus Biologicals™
Rabbit Monoclonal Antibody
286.00€ - 581.00€
Specifications
| Antigen | Histone H2a |
|---|---|
| Clone | S08-1A5 |
| Dilution | Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 1:10-1:500, Immunofluorescence |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18383036
|
Novus Biologicals
NBP3-15040-25UL |
25 μg |
286.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18396953
|
Novus Biologicals
NBP3-15040-100UL |
100 μg |
581.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Histone H2a Monoclonal antibody specifically detects Histone H2a in Yeast samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ImmunofluorescenceSpecifications
| Histone H2a | |
| Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 1:10-1:500, Immunofluorescence | |
| Monoclonal | |
| Purified | |
| RUO | |
| Yeast | |
| H2A histone family, member Q, H2A/q, H2AFQH2A, H2A-GL101, histone 2, H2ac, histone cluster 2, H2ac, histone H2A type 2-C, Histone H2A/q, Histone H2A-GL101, histone IIa, MGC74460 | |
| A synthetic peptide of Histone H2a (Uniprot#: P04912) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| S08-1A5 | |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Chromatin Research, Epigenetics | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
| 8338 | |
| IgG | |
| Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title