missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIP2 Monoclonal Antibody (701316)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561693
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
Accepts ubiquitin from the E1 complex and catalyzes its covalent attachment to other proteins. In vitro, in the presence or in the absence of BRCA1-BARD1 E3 ubiquitin-protein ligase complex, catalyzes the synthesis of 'Lys-48'-linked polyubiquitin chains. Does not transfer ubiquitin directly to but elongates monoubiquitinated substrate protein. Mediates the selective degradation of short-lived and abnormal proteins, such as the endoplasmic reticulum-associated degradation (ERAD) of misfolded lumenal proteins. Ubiquitinates huntingtin. May mediate foam cell formation by the suppression of apoptosis of lipid-bearing macrophages through ubiquitination and subsequence degradation of p53/TP53. Proposed to be involved in ubiquitination and proteolytic processing of NF-kappa-B; in vitro supports ubiquitination of NFKB1. In case of infection by cytomegaloviruses may be involved in the US11-dependent degradation of MHC class I heavy chains following their export from the ER to the cytosol. In case of viral infections may be involved in the HPV E7 protein-dependent degradation of RB1.
Specifications
| HIP2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P61086, P61087 | |
| UBE2K | |
| E. coli-derived recombinant human UBE2K/E2-25K, Ala2-Asn200. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| 701316 | |
| Unconjugated | |
| UBE2K | |
| AW492011; D5Ertd601e; DKFZp564C1216; DKFZp686J24237; E2 ubiquitin-conjugating enzyme K; E2(25K); E2-25k; HIP2; HIP-2; huntingtin interacting protein 2; huntingtin-interacting protein 2; HYPG; LIG; UBC1; UBE2K; Ubiquitin carrier protein; ubiquitin conjugating enzyme E2 K; ubiquitin conjugating enzyme E2K; ubiquitin-conjugating enzyme E2 K; Ubiquitin-conjugating enzyme E2(25K); ubiquitin-conjugating enzyme E2-25 KDA; ubiquitin-conjugating enzyme E2-25K; ubiquitin-conjugating enzyme E2K; ubiquitin-conjugating enzyme E2K (UBC1 homolog, yeast); ubiquitin-protein ligase | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 3093, 53323 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction