missing translation for 'onlineSavingsMsg'
Learn More

HIP-55 Antibody (OTI6C9), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18162152 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18162152 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18162152 Supplier Novus Biologicals Supplier No. NBP271841AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HIP-55 Monoclonal specifically detects HIP-55 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIP-55
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI6C9
Conjugate Alexa Fluor 750
Dilution Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias ABP1, Cervical SH3P7, CMAP, Drebrin-F, drebrin-like, drebrin-like protein, HIP-55SH3P7Cervical mucin-associated protein, HPK1-interacting protein of 55 kDa, SH3 domain-containing protein 7, src homology 3 domain-containing protein HIP-55
Gene Symbols DBNL
Host Species Mouse
Immunogen Full length recombinant protein of human HIP-55 (NP_054782) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 28988
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.