Learn More
HINT1 Antibody, Novus Biologicals™
Product Code. 18707403 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.10mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18707403 0.1 mL 0.10mL
18446540 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18707403 Supplier Novus Biologicals Supplier No. NBP183257
Rabbit Polyclonal Antibody
HINT1 Polyclonal specifically detects HINT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Applications Western Blot, Immunohistochemistry
Host Species Rabbit
Quantity 0.1 mL