missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEY1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
483.00€
Specifications
| Antigen | HEY1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
HEY1 Polyclonal specifically detects HEY1 in Rat samples. It is validated for Western Blot.Specifications
| HEY1 | |
| Polyclonal | |
| Rabbit | |
| NP_001178774 | |
| 23462 | |
| Synthetic peptide towards Hey1. Peptide sequence WGSAFGHHPHIAHPLLLPQNGHGNAGTTASPTEPHHQGRLASAHPEAPAL. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| basic helix-loop-helix protein OAF1, BHLHb31, Cardiovascular helix-loop-helix factor 2, CHF-2, CHF2hHRT1, Class B basic helix-loop-helix protein 31, Hairy and enhancer of split-related protein 1, hairy/enhancer-of-split related with YRPW motif 1, hairy/enhancer-of-split related with YRPW motif protein 1, Hairy-related transcription factor 1, HERP2HES-related repressor protein 1, HESR-1, HESR1MGC1274, HES-related repressor protein 2, HRT1, HRT-1OAF1 | |
| HEY1 | |
| IgG | |
| 32 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title