missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexokinase 2 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
213.00€ - 507.00€
Specifications
| Antigen | Hexokinase 2 |
|---|---|
| Dilution | Western Blot 1:5000 - 1:30000, ELISA Recommended starting concentration is 1 μg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30231457
|
Novus Biologicals
NBP3-33349-100ul |
100 μL |
507.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30229002
|
Novus Biologicals
NBP3-33349-20ul |
20 μL |
213.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Hexokinase 2 Monoclonal antibody specifically detects Hexokinase 2 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| Hexokinase 2 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 3099 | |
| IgG | |
| Affinity purified |
| Western Blot 1:5000 - 1:30000, ELISA Recommended starting concentration is 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DKFZp686M1669, EC 2.7.1, hexokinase 2, Hexokinase type II, HKII, HXK2, muscle, Muscle form hexokinase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human Hexokinase 2 (NP_000180.2).,, Sequence:, MIASHLLAYFFTELNHDQVQKVDQYLYHMRLSDETLLEISKRFRKEMEKGLGATTHPTAAVKMLPTFVRSTPDGTEHGEFLALDLGGTNFRVLWVKVTDNGLQKVEMENQIYAIPEDIMR | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title